ENA394767930 Enamine

2-(pyrrolidin-3-yl)-2H-indazole

  • MDL number MFCD24843010
  • Cas Number : 1443979-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129981
Availability details for
Quantity
  • Availability
  • Add to Cart