ENA394767932 Enamine

5-methyl-4,5-dihydro-1H-imidazol-2-amine hydrobromide

  • MDL number MFCD24843014
  • Cas Number : 1443981-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130020

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-130020
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart