ENA394767935 Enamine

tert-butyl N-[1-(cyclohexylmethyl)-1H-1,2,3-triazol-4-yl]carbamate

  • MDL number MFCD24843017
  • Cas Number : 1443981-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130075
Availability details for
Quantity
  • Availability
  • Add to Cart