ENA394767943 Enamine

tert-butyl N-[2-(prop-2-yn-1-ylsulfanyl)ethyl]carbamate

  • MDL number MFCD24394513
  • Cas Number : 1058159-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130111
Availability details for
Quantity
  • Availability
  • Add to Cart