ENA394767944 Enamine

2-cyano-3-(thiophen-3-yl)propanamide

  • MDL number MFCD24843026
  • Cas Number : 1443981-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130135

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130135
Availability details for
Quantity
  • Availability
  • Add to Cart