ENA394767945 Enamine

cycloheptylmethyl methanesulfonate

  • MDL number MFCD24843027
  • Cas Number : 75646-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130145
Availability details for
Quantity
  • Availability
  • Add to Cart