ENA394767950 Enamine

1-(3,4,5-trifluorophenyl)ethane-1,2-diamine dihydrochloride

  • MDL number MFCD24843032
  • Cas Number : 1443979-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130194
Availability details for
Quantity
  • Availability
  • Add to Cart