ENA394767963 Enamine

6-(aminomethyl)-2-ethylpyridine-3-carboxylic acid dihydrochloride

  • MDL number MFCD24843048
  • Cas Number : 1443981-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130306
Availability details for
Quantity
  • Availability
  • Add to Cart