ENA394767965 Enamine

1H,2H,3H-pyrrolo[3,2-b]pyridine dihydrochloride

  • MDL number MFCD24843050
  • Cas Number : 1443981-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130308
Availability details for
Quantity
  • Availability
  • Add to Cart