ENA394767970 Enamine

1-(5-methyl-1,3,4-oxadiazol-2-yl)ethyl acetate

  • MDL number MFCD24843055
  • Cas Number : 1443981-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130366
Availability details for
Quantity
  • Availability
  • Add to Cart