ENA394767971 Enamine

tert-butyl N-{2-[(cyclobutylmethyl)sulfanyl]ethyl}carbamate

  • MDL number MFCD24843056
  • Cas Number : 1443979-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130379
Availability details for
Quantity
  • Availability
  • Add to Cart