ENA394767985 Enamine

3-methyl-1-[3-(methylamino)propyl]-1,2-dihydroquinoxalin-2-one hydrochloride

  • MDL number MFCD24843072
  • Cas Number : 1443979-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130923
Availability details for
Quantity
  • Availability
  • Add to Cart