ENA394767987 Enamine

2-chloro-N-{10,14-dioxa-4-thia-6-azatricyclo[7.5.0.0,3,7]tetradeca-1,3(7),5,8-tetraen-5-yl}acetamide

  • MDL number MFCD24843074
  • Cas Number : 1443979-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-46160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46160
Availability details for
Quantity
  • Availability
  • Add to Cart