ENA394767988 Enamine

methyl 2-(2-chloroacetamido)-2-methylpentanoate

  • MDL number MFCD24843075
  • Cas Number : 1443980-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-47300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47300
Availability details for
Quantity
  • Availability
  • Add to Cart