ENA394767998 Enamine

N-[4-chloro-3-(trifluoromethyl)phenyl]-2,2-dimethylpropanamide

  • MDL number MFCD02073387
  • Cas Number : 13691-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129514
Availability details for
Quantity
  • Availability
  • Add to Cart