ENA394767999 Enamine

6-(dimethylamino)hexan-2-one

  • MDL number MFCD12058287
  • Cas Number : 4305-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129810
Availability details for
Quantity
  • Availability
  • Add to Cart