ENA394768013 Enamine

3-amino-1-[2-(morpholin-4-yl)ethyl]piperidin-2-one

  • MDL number MFCD19679231
  • Cas Number : 1342059-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129828
Availability details for
Quantity
  • Availability
  • Add to Cart