ENA394768014 Enamine

[(4-bromophenyl)methyl]thiourea

  • MDL number MFCD14648215
  • Cas Number : 89980-86-9
  • Molecular Weight : 245.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129829
Availability details for
Quantity
  • Availability
  • Add to Cart