ENA394768024 Enamine

5-iodoquinazolin-4-ol

  • MDL number MFCD15527378
  • Cas Number : 860193-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129523
Availability details for
Quantity
  • Availability
  • Add to Cart