ENA394768031 Enamine

2,3-dichloro-4-fluorobenzonitrile

  • MDL number MFCD19690560
  • Cas Number : 908123-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129933
Availability details for
Quantity
  • Availability
  • Add to Cart