ENA394768037 Enamine

methyl 2-amino-4-fluoro-3-methylbenzoate

  • MDL number MFCD16066175
  • Cas Number : 928839-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35...37
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129941

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129941
Availability details for
Quantity
  • Availability
  • Add to Cart