ENA394768039 Enamine

4-chloro-5-iodoquinazoline

  • MDL number MFCD09954864
  • Cas Number : 959237-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129944
Availability details for
Quantity
  • Availability
  • Add to Cart