ENA394768046 Enamine

2-bromo-3,3-dimethylbutanamide

  • MDL number MFCD00068787
  • Cas Number : 25117-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129973

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129973
Availability details for
Quantity
  • Availability
  • Add to Cart