ENA394768048 Enamine

1-bromo-3-[(2,2,2-trifluoroethoxy)methyl]benzene

  • MDL number MFCD14686963
  • Cas Number : 1248738-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129975
Availability details for
Quantity
  • Availability
  • Add to Cart