ENA394768050 Enamine

methyl 3-(bromomethyl)-4-fluorobenzoate

  • MDL number MFCD16036856
  • Cas Number : 878744-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129978

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129978
Availability details for
Quantity
  • Availability
  • Add to Cart