ENA394768057 Enamine

benzyl 1H-indole-5-carboxylate

  • MDL number MFCD13176765
  • Cas Number : 136564-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129986

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129986
Availability details for
Quantity
  • Availability
  • Add to Cart