ENA394768064 Enamine

2-chloro-4-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)aniline

  • MDL number MFCD20296392
  • Cas Number : 1339515-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130004
Availability details for
Quantity
  • Availability
  • Add to Cart