ENA394768069 Enamine

2-(prop-2-en-1-ylsulfanyl)ethan-1-amine

  • MDL number MFCD09944195
  • Cas Number : 42416-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130018
Availability details for
Quantity
  • Availability
  • Add to Cart