ENA394768071 Enamine

1-(benzylsulfanyl)-2-(chloromethyl)benzene

  • MDL number MFCD24842984
  • Cas Number : 4521-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129483
Availability details for
Quantity
  • Availability
  • Add to Cart