ENA394768076 Enamine

2-(benzylsulfanyl)acetonitrile

  • MDL number MFCD00019834
  • Cas Number : 17377-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130065
Availability details for
Quantity
  • Availability
  • Add to Cart