ENA394768078 Enamine

1-(piperazin-1-yl)-2-(pyridin-4-yl)ethan-1-one

  • MDL number MFCD09729568
  • Cas Number : 849355-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129644
Availability details for
Quantity
  • Availability
  • Add to Cart