ENA394768079 Enamine

4-(1-aminoethyl)-2-methylbenzonitrile

  • MDL number MFCD18706389
  • Cas Number : 1270420-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129646
Availability details for
Quantity
  • Availability
  • Add to Cart