ENA394768083 Enamine

ethyl 4-(trifluoromethyl)-1H-pyrrole-3-carboxylate

  • MDL number MFCD09838835
  • Cas Number : 120732-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-164
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129487
Availability details for
Quantity
  • Availability
  • Add to Cart