ENA394768087 Enamine

2-bromo-1-(3,4-difluorophenyl)-2-methylpropan-1-one

  • MDL number MFCD19304158
  • Cas Number : 1310094-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130108
Availability details for
Quantity
  • Availability
  • Add to Cart