ENA394768088 Enamine

1-(3-chloro-2-fluorophenyl)-2-methoxyethan-1-one

  • MDL number MFCD21953690
  • Cas Number : 1443981-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130109
Availability details for
Quantity
  • Availability
  • Add to Cart