ENA394768098 Enamine

3-amino-3-(2-bromophenyl)propan-1-ol

  • MDL number MFCD06227470
  • Cas Number : 171911-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130130

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130130
Availability details for
Quantity
  • Availability
  • Add to Cart