ENA394768103 Enamine

7-chloro-1-(2-phenoxyethyl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD24842990
  • Cas Number : 1303832-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129659
Availability details for
Quantity
  • Availability
  • Add to Cart