ENA394768112 Enamine

N-ethyl-2,5-difluorobenzene-1-sulfonamide

  • MDL number MFCD05086730
  • Cas Number : 898074-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130157
Availability details for
Quantity
  • Availability
  • Add to Cart