ENA394768117 Enamine

3-(3-methoxybenzoyl)-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD11607105
  • Cas Number : 1082867-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...129
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130173

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130173
Availability details for
Quantity
  • Availability
  • Add to Cart