ENA394768119 Enamine

1-(4-cyclopropyl-1,3-thiazol-2-yl)ethan-1-ol

  • MDL number MFCD19607004
  • Cas Number : 1343645-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130190
Availability details for
Quantity
  • Availability
  • Add to Cart