ENA394768121 Enamine

1-benzyl-3,3-dimethylazetidine

  • MDL number MFCD21338041
  • Cas Number : 13509-71-2
  • Molecular Weight : 175.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl-3,3-dimethylazetidine
Inchi Code 1S/C12H17N/c1-12(2)9-13(10-12)8-11-6-4-3-5-7-11/h3-7H,8-10H2,1-2H3
Inchi Key WPDLZIHBUNVNLF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129665
Availability details for
Quantity
  • Availability
  • Add to Cart