ENA394768127 Enamine

5-benzyl-1,3-oxazolidin-2-one

  • MDL number MFCD01755618
  • Cas Number : 42746-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130206

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130206
Availability details for
Quantity
  • Availability
  • Add to Cart