ENA394768128 Enamine

4-benzyl-2-(5-methyl-1,2,4-oxadiazol-3-yl)morpholine

  • MDL number MFCD24843034
  • Cas Number : 1381581-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130219
Availability details for
Quantity
  • Availability
  • Add to Cart