ENA394768132 Enamine

dimethyl(pyrrolidin-3-ylmethyl)amine hydrochloride

  • MDL number MFCD22121387
  • Cas Number : 1443980-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129669

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129669
Availability details for
Quantity
  • Availability
  • Add to Cart