ENA394768140 Enamine

tert-butyl 3-(bromomethyl)benzoate

  • MDL number MFCD16620833
  • Cas Number : 126062-63-3
  • Molecular Weight : 271.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-130245

Safety Information

signalword Danger
pcodes P234,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P390,P405,P406,P501
MSDS https://www.enaminestore.com/catalog/EN300-130245
pictograms GHS05
hstatements H290,H314
Availability details for
Quantity
  • Availability
  • Add to Cart