ENA394768141 Enamine

5-fluoro-1-[2-(3-methylphenoxy)ethyl]-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD24843040
  • Cas Number : 1291543-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130247
Availability details for
Quantity
  • Availability
  • Add to Cart