ENA394768142 Enamine

4-fluoro-N1-[2-(3-methylphenoxy)ethyl]benzene-1,2-diamine

  • MDL number MFCD16284571
  • Cas Number : 1270795-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130248
Availability details for
Quantity
  • Availability
  • Add to Cart