ENA394768147 Enamine

8-fluoro-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD19285320
  • Cas Number : 127557-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130254
Availability details for
Quantity
  • Availability
  • Add to Cart