ENA394768150 Enamine

3-methyl-1-phenyl-1H-pyrazol-4-amine

  • MDL number MFCD20691037
  • Cas Number : 103095-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-90
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130259
Availability details for
Quantity
  • Availability
  • Add to Cart