ENA394768151 Enamine

7-bromo-5-methyl-1H,2H,3H,4H,9H-cyclopenta[b]quinolin-9-one

  • MDL number MFCD14653704
  • Cas Number : 1247186-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130260

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130260
Availability details for
Quantity
  • Availability
  • Add to Cart